Would you like to log in again?
SNAP-CELL® 505-Star, New England Biolabs
Supplier: New England Biolabs
S9103S S9103S
101447-656EA 396 USD InStock
CA101447-656 101447-656
SNAP-CELL® 505-Star, New England Biolabs
Protein Linking and Labeling Systems
SNAP-Cell® 505-Star is a photostable green fluorescent substrate that can be used to label SNAP-tag® fusion proteins inside living cells, on cell surfaces, or in vitro
This cell-permeable substrate (CP-6-505) is based on the single isomer 6-carboxyrhodamine 110 and is suitable for standard fluorescein filter sets. It has an excitation maximum at 504 nm and an emission maximum at 532 nm. This package contains 50 nmol of SNAP-Cell 505-Star substrate, sufficient to make 10 ml of a 5 µM SNAP-tag fusion protein labeling solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty