Would you like to log in again?
SNAP-CELL® TMR-STAR , New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
S9105S S9105S
101447-660EA 396 USD InStock
101447-660 CA101447-660
SNAP-CELL® TMR-STAR , New England Biolabs
Protein Linking and Labeling Systems
SNAP-Cell® TMR-Star is a red fluorescent substrate that can be used to label SNAP-tag® fusion proteins inside living cells, on cell surfaces, or in vitro
This cell-permeable substrate (CP-6-TMR) is based on the single isomer 6-carboxytetramethylrhodamine and is suitable for standard rhodamine filter sets. It has an excitation maximum at 554 nm and an emission maximum at 580 nm. This package contains 30 nmol of SNAP-Cell TMR-Star substrate, sufficient to make 10 ml of a 3 µM SNAP-tag fusion protein labeling solution.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty