Would you like to log in again?
BG-PEG-NH2, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
S9150S S9150S
101447-708EA 466 USD InStock
CA101447-708 101447-708
BG-PEG-NH2, New England Biolabs
Protein Linking and Labeling Systems
BG-PEG-NH2 is an amine-terminated building block for the one-step synthesis of SNAP-tag substrates from NHS esters or other activated carboxyl esters on labels or surfaces
This compound allows you to make custom SNAP-tag substrates for labeling SNAP-tag fusion proteins for a wide range of applications. BG-PEG-NH2 is particularly useful for the synthesis of SNAP-tag substrates with affinity ligands or solid surfaces.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty