Would you like to log in again?
CLIP-CELL® TMR-Star, New England Biolabs
Supplier: New England Biolabs
S9219S S9219S
101447-722EA 396 USD InStock
CA101447-722 101447-722
CLIP-CELL® TMR-Star, New England Biolabs
Protein Linking and Labeling Systems
CLIP-Cell™ TMR-Star is a photostable red fluorescent substrate that can be used to label CLIP-tag™ fusion proteins inside living cells, on cell surfaces or in vitro
This cell-permeable substrate (BC-TMR) is based on tetramethylrhodamine and is suitable for standard rhodamine filter sets. It has an excitation maximum at 554 nm and emission maximum at 580 nm. This package includes 30 nmol of CLIP-Cell TMR-Star, sufficient to make 10 ml of a 3 µM CLIP-tag fusion protein labeling solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty