Would you like to log in again?
CLIP-Biotin, New England Biolabs
Supplier: New England Biolabs
S9221S S9221S
101447-726EA 315 USD InStock
CA101447-726 101447-726
CLIP-Biotin, New England Biolabs
Protein Linking and Labeling Systems
CLIP-Biotin is a cell-permeable substrate (BC-Biotin) based on biotin with an amidocaproyl linker
It is suitable for applications such as biotinylation of CLIP-tag™ fusion proteins in living cells for detection with streptavidin fluorophore conjugates or labeling in solution for analysis by SDS-PAGE/Western Blot or for capture with streptavidin for binding and interaction studies. This package contains 50 nmol of CLIP-Biotin substrate, sufficient to make 10 ml of a 5 μM solution for the labeling of CLIP-tag fusion proteins in cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty