Would you like to log in again?
Tma Endonuclease III
Supplier: New England Biolabs
M0291S M0291S
101449-296EA 85 USD InStock
CA101449-296 101449-296
Tma Endonuclease III
Enzymes
A thermostable homolog of the E.coli Endonuclease III (Nth). It acts as an N-glycosylase and an AP-lyase.
The N-glycosylase activity releases damaged pyrimidines from double-stranded DNA, generating an apurinic (AP site). The AP-lysase activity then cleaves the resulting abasic site. Tma Endonuclease III recognizes abasic sites, 5,6 dihydroxythymine and thymine glycol in DNA.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty