Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hei-VAP Industrial Rotary Evaporators, 220 V

Supplier: Heidolph
036072055 036072060 036072075 036072050 036072065 036072095 036072090 036072070 036072065 036072080 036072085
10145-526EA 66003.17 USD InStock
10145-526 10145-514 10145-524 CA10145-520 10145-522 10145-532 10145-520 10145-530 10145-518 10145-528 10145-516
Hei-VAP Industrial Rotary Evaporators, 220 V
Evaporators Rotary Evaporators
Designed for advanced requirements and integrated vacuum control capabilities

  • 220 V model

Units feature a large touch screen control panel with illuminated displays for all process parameters and programmable ramps. Universal heating bath accommodates water or other bath fluids allowing for tempearture setting up to 180 °C (356 °F).

Optional base cart is fully maneuverable with removable panels for storage of vacuum pump, glassware, solvents, or accessories.

Evaporators come standard with integrated refill water system, spillover preventions, and a release valve on the bottom.

Certifications: Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ).

Ordering information: Glass set included. Base and all other accessories not included.

Caution: 220 V model.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty