Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sample Cylinders, Ultra-High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Supplier: Restek
22929 22927-PI 22111 22113-PI 22111-PI 22928-PI 22112-PI 22928 22929-PI 22113 22112 22927
10145-834EA 966.15 USD InStock
10145-834 10145-734 10145-842 10145-732 10145-840 10145-730 10145-740 10145-728 10145-838 10145-836 10145-726 10145-736
Sample Cylinders, Ultra-High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Cylinders Hydrometer Cylinders
Swagelok® sample cylinders are made of 304L and 316L stainless steel to resist corrosion and DOT rated to 1,800 and 5,000 psig (TPED cylinders rated to 1,450 and 4,350 psig), which allows sampling at gas wellheads as well as on-site refineries.
  • 316L stainless steel; DOT rating to 5,000 psig (TPED cylinders to 4,350 psig)
  • Range of cylinder sizes, 150 cc to 500 cc
  • All cylinders have 1/4" female NPT threads on both ends
  • TPED compliant cylinders available for EU community
Each cylinder is hydrostatically tested to at least 5/3 the working pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty