Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sample Cylinders, High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Supplier: Restek
24132 24130 24132-PI 21394-PI 22926-PI 22925-PI 24131-PI 22923 22926 22922-PI 22922 22921-PI 24133 22924 24134-PI 22924-PI 22923-PI 22925 24133-PI 22921 24134 21394 24131 24130-PI
10145-746EA 481.99 USD InStock
10145-746 10145-812 10145-844 10145-822 10145-832 10145-754 10145-820 10145-830 10145-852 10145-752 10145-850 10145-750 10145-682 10145-680 10145-818 10145-828 10145-816 10145-826 10145-848 10145-738 10145-814 10145-748 10145-846 10145-824
Sample Cylinders, High Pressure (Stainless Steel & Sulfinert®-Treated), Restek
Cylinders Hydrometer Cylinders
Swagelok® sample cylinders are made of 304L and 316L stainless steel to resist corrosion and are DOT rated to 1,800 and 5,000 psig (TPED cylinders rated to 1,450 and 4,350 psig), which allows sampling at gas wellheads as well as on-site refineries.
  • 304L stainless steel; DOT rating to 1,800 psig (TPED cylinders to 1,450 psig)
  • Range of cylinder sizes, from 75 cc to 2,250 cc
  • All cylinders have 1/4" female NPT threads on both ends
  • TPED compliant cylinders available for EU community
Each cylinder is hydrostatically tested to at least 5/3 the working pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty