Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sample Cylinder Carrying Handle, Restek
Supplier: Restek
26373 26374
10145-940EA 120.78 USD InStock
10145-940 10145-938
Sample Cylinder Carrying Handle, Restek
Cylinders Hydrometer Cylinders
304 stainless steel fabrication provides a sturdy, durable handle for easy carrying of sample cylinders.
  • Stainless steel fabrication
  • Compatible with 1.9" and 2" OD cylinders
Includes a stainless steel handle and two attachment rings for holding sample cylinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty