Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sample Cylinder 316 SS End Pipe Plug, Restek
Supplier: Restek
26375 26376
10145-944EA 68.44 USD InStock
10145-944 10145-942
Sample Cylinder 316 SS End Pipe Plug, Restek
Cylinders Hydrometer Cylinders
Sample cylinder plugs protect Swagelok tube fitting or NPT end connections on valves during cylinder transport.
  • Stainless steel fabrication
Fitting type: 1/4" male NPT
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty