Would you like to log in again?
Microtubes without Cap, Sarstedt
Supplier: Sarstedt
72.708 72.696
101453-235PK 48.74 USD InStock
101453-235 101093-233
Microtubes without Cap, Sarstedt
Tubes Microtubes
Transparent polypropylene microtubes are supplied without caps.
Microtubes can withstand centrifugation up to 20000 ×g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty