Would you like to log in again?
Sample Stub Vacuum Desiccators , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76550
101454-046EA 1271.46 USD InStock
101454-046
Sample Stub Vacuum Desiccators , Electron Microscopy Sciences
Desiccators Desiccator Cabinets
Precision machined from a block of aluminum – anodized black - this Sample Stub Vacuum Desiccators accommodates 18 SEM Mounted Sample Stubs, pin style, under vacuum for year long
13 x 13 x 4.5 cm. Chamber cavity is 10.4 cm diameter and 18 mm deep. Lid is made with 10 mm thick clear acrylic for easyviewing the content. Prevents oxidation from occurring  to the sample during long term storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty