Would you like to log in again?
TorrLube® Vacuum Oil and Lubricant, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60750-01 60750-20
101454-048EA 194.37 USD InStock
101454-048 101454-050
TorrLube® Vacuum Oil and Lubricant, Electron Microscopy Sciences
Lubricants
TorrLube® Oil is an extremely pure PFPE (Perfluoropolyalkether) blend which is thoroughly distilled and then filtered through a submicron screen
These properties make it the ideal lubricant for bearings, automated slides, and screws that have a tendency to gall or cold weld while working under extreme conditions. TorrLube® Extreme Grease is white, odorless, nonflammable, and chemically inert products based on TorrLube® Oil and the most advanced PTFE thickener available.
Each syringe contains 1 cc. of TorrLube® lubricant with a #22 gauge blunt needle attached for close application. Syringes are packaged in a clean room environment and the needles are sealed to prevent contaminant or leakage.
It has the ability to withstand thermal expansion and contraction as well as chemical and mechanical stress, while still maintaining consistent viscosity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty