Would you like to log in again?
Polyethylene Molds, Individual Molds, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70177-12 70177-16 70177-19 70178-04
101454-086PK 272.85 USD InStock
101454-086 101454-084 101454-088 101454-082
Polyethylene Molds, Individual Molds, Electron Microscopy Sciences
Moulds Moulds for Histology
Molded from polyethylene, three popular sizes are available to meet your needs.
These Individual Molds can be used with EBH2 Block Holders, Aluminum Embedding Tray is available with 6-position to hold these Individual Mold during polymerization. Comes with anodized blue and natural color. Suitable for JB-4®, JB-4-Plus®, Immuno-Bed, LR White, LR Gold® or Paraffin embedding.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty