Would you like to log in again?
Microtube Rack - Microwaveable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72372-R20 72372-SB 72372-SW
101454-124EA 56.23 USD InStock
101454-124 101454-122 101454-118
Microtube Rack - Microwaveable, Electron Microscopy Sciences
Racks Tube Racks
Polypropylene floating racks keep microtubes submerged in a water bath – perfect for controlling sample temperature. Detachable legs give the users more options: attach legs so rack stands upright on benchtop, or detach legs so sample tubes can be quickly popped out by pushing down onto any surface. Ideally to hold microcentrifuge tube in water bath during polymerization, such as LR White®. Not autoclavable. Measures: 60 mm high including legs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty