Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Media Disruptor Genie, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72406-10 72407-EU 72407-10 72406-AU 72406-12 72407-BR 72408-01 72407-AU 72406-EU 72406-20 72406-BR 72408-05 72407-SW 72407-12 72406-SW 72407-20
101454-142EA 1441.54 USD InStock
101454-142 101454-130 101454-152 101454-140 101454-150 101454-146 101454-134 101454-156 101454-144 101454-132 101454-154 101454-128 101454-138 101454-126 101454-148 101454-136
Media Disruptor Genie, Electron Microscopy Sciences
Homogenizers Ultrasonic Cell Disruptors
Rapid hands-free disruption of up to 12 microtubes (1.5 ml or 2.0 ml) at one time.
  • Unique agitation, vortexing, and collision actions for cell disruption and sample resuspension
  • Maintain operating speed for up to 15 minutes or continuous operation, even in cold rooms
  • Supplied pop-up cup easily attaches for use as a standard single-tube vortex mixer
  • Ideal for glass bead procedures using Disruptor Beads
  • Less costly than ultrasonic units or homogenizes
  • Spherecal lead-free soda lime glass disruptor beads are available in two sizes
  • Beads can be used for disruption of yeast, bacteria, and other samples
  • Beads can be used in combination with Disruptor Genie or TurboMix™ attachments for Vortex-Genie™ 2, Vortec-Genie™ 2T or Digital Vortex-Genie™ 2
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty