Would you like to log in again?
Cleanroom Wipes for Grab-EEZ™ Wipe Dispenser
Supplier: High-Tech Conversions
GE-NT1-712
10146-288CS 420.92 USD InStock
10146-288
Cleanroom Wipes for Grab-EEZ™ Wipe Dispenser
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Nonwoven poly-cellulose cleanroom wipes for use with the Grab-EEZ™ wipe dispenser.
  • Double C-folding allows you to easily grab only one wipe
  • Extremely low in particles
ISO Class 5 (Class 100) cleanroom compatible.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty