Would you like to log in again?
Straight Side Glass Bottles, Clear, C&G Containers
Supplier: C&G Containers
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LFS002100000 LFS002200000 LFS004200100 LFS002300000 LFS008100000 LFS008100010 LFS032230010 LFS032900800 LFS004400100 LFS004300000 LFS016400000 LFS002400010 LFS016900800 LFS016100000 LFS002900000 LFS032200000 LFS002900500 LFS002400000 LFS016200000 LFS002400410 LFS016200010 LFS032100010 LFS008200010 LFS002200010 LFS032200010 LFS002100010 LFS004900100 LFS004900500 LFS004100010 LFS008400010 LFS016900100 LFS004200000 LFS032200000 LFS004100000 LFS0322100 LFS032900000 LFS004900000 LFS004400010 LFS008900000
89125-404CS 114.5 USD InStock
89125-404 71000-852 71000-830 71000-850 10146-730 30621-200 30621-188 71000-846 71000-824 10146-770 30621-202 71000-822 71000-844 71000-826 71000-848 30621-204 71000-842 71000-820 71000-840 10496-904 30617-864 10146-758 10496-908 71000-818 30621-194 10146-760 10496-910 30621-190 10496-912 71000-834 71000-812 71000-832 30621-196 71000-816 71000-838 89125-408 30621-198 71000-814 71000-836
Straight Side Glass Bottles, Clear, C&G Containers
Jars General Use Jars
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles, Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some bottles are chemically preserved.
Secondary uses include containment, bench vessels, and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity "contain" volumes may vary.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty