Would you like to log in again?
Cryobox for PolarSafe™ Cryogenic Storage Vials with Star Caps, Argos Technologies
Supplier: Argos Technologies
04395-87 04395-84 4395-85 04395-86
10146-830EA 45.03 USD InStock
10146-830 10146-800 10146-832 10146-828
Cryobox for PolarSafe™ Cryogenic Storage Vials with Star Caps, Argos Technologies
Boxes Cryoboxes
The Argos PolarSafe polycarbonate storage boxes work in conjuciton with the 2D tubes to provide automatic identification of both the box and stored vials.
  • Each box features a 2D Datamatrix code on the bottom of the box as well as a laser engraved Code 128 barcode on the side Offered in both 81 and 100-well formats
PolarSafe 2D boxes are available in “short” (for 2 ml tubes) and “tall” designs (for 5 ml tubes).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty