Would you like to log in again?
Bolton Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
U83
10146-804PK 598.73 USD InStock
10146-804
Bolton Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Campylobacter spp
For the selective enrichment of Campylobacter spp. from food and dairy products.
from food and dairy products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty