Would you like to log in again?
R2A Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
U354
10146-856PK 339.5 USD InStock
10146-856
R2A Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the detection of heterotrophic bacteria in water, by membrane filtration technique.
  • Membrane filtration technique
For the detection of heterotrophic bacteria in water, by membrane filtration technique.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty