Would you like to log in again?
hema-screen™, Immunostics
Supplier: Immunostics
Hema-Screen®
HS-34 HS-100 HSTR-335CS HS-50 HS-1000 HSPP-50
10147-014KT 72.1 USD InStock
10147-014 10147-024 10147-011 10147-028 10147-020 10147-030
hema-screen™, Immunostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
hema-screen™ is a rapid, convenient, and non-offensive qualitative method for detecting occult blood in the stool
  • Detect hemoglobin as low as 0.6 mg Hb/gm of feces
  • Test time within 30 seconds
Recommended for routine hospital testing and mass screening programs for colorectal cancer detection.
Certifications: CLIA Waived. CE Marked.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty