Would you like to log in again?
hema-screen™ SPECIFIC, Fecal Immunochemical Occult Blood Test, Immunostics
Supplier: Immunostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Hema-Screen®
HSSPENV-20 HSSP-25 HSSPCON HSSPCAS-25
10147-034KT 285.29 USD InStock
10147-034 10147-038 10147-032 10147-040
hema-screen™ SPECIFIC, Fecal Immunochemical Occult Blood Test, Immunostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
hema-screen SPECIFIC is a rapid fecal immunochemical test (FIT)
This test delivers a convenient, qualitative immunoassay for the determination of human hemoglobin in feces. It is a vital tool in the diagnosis and therapy of gastrointestinal disorders. hema-screen SPECIFIC features an innovative sampling method that utilizes one card for the collection of two specimens of feces and only one immunochemical specimen preparation tube or test cassette system for analysis.
Ordering information: The hema-screen SPECIFIC Kit comes with 25 cassettes, 25 buffer tubes, 35 mailing envelopes, 1 product insert and 1 procedure card. The mailing envelopes contain 1 DEVEL-A-TAB slide, 2 collection tissues, 2 applicator sticks and patient instructions.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty