Would you like to log in again?
AzureRed Fluorescent Protein Stain, Azure BioSystems
Supplier: Azure Biosystems
AC2124
10147-342EA 316.73 USD InStock
10147-342
AzureRed Fluorescent Protein Stain, Azure BioSystems
Assays Protein Assays
AzureRed Fluorescent Protein Stain is a quantitative total protein stain for blots and blots with the sensitivity to detect less than 1 ng of protein per band or spot
Gels and blots stained with AzureRed can be imaged on any fluorescent imaging system, including laser- and CCD-based systems.
Fully compatible with downstream Western blotting or mass spectrometry, AzureRed is non-toxic and biodegradable, for safe and simple disposal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty