Would you like to log in again?
AzureSpectra Fluorescent Western Blotting Kits, Azure Biosystems
Supplier: Azure Biosystems
AC2104 AC2140 AC2141
10147-376KT 444.41 USD InStock
10147-376 10147-374 10147-378
AzureSpectra Fluorescent Western Blotting Kits, Azure Biosystems
Blotting Kits
AzureSpectra Kits provide everything you need to conduct two-color fluorescent Western blots.
Each kit includes pre-cut membranes, wash buffer, blocking buffer, quenching sheets, and secondary antibodies to detect two proteins per experiment.
Choose from a selection of kits for visible or NIR fluorescent imaging. Demo kits include reagents, including protein samples and transfer membrane, to conduct one sample blot.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty