Would you like to log in again?
SP Bel-Art Secador® 4.0 Horizontal Profile Gas-Purge Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
F42074-0002
10147-470EA 1323.35 USD InStock
10147-470
SP Bel-Art Secador® 4.0 Horizontal Profile Gas-Purge Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
This sturdy Secador® desiccator cabinet has a large, wide storage space and provides convenient, reliable dry storage of moisture- and UV-sensitive products.
  • Convenient, reliable dry storage
  • Protects moisture- and UV-sensitive products
  • Co-polyester construction
  • Tapered rear-mounted gas ports
Durastar® co-polyester construction blocks 99% of UV light and is resistant to staining, crazing and chemical attack
Stackable up to 2 units high with use of Stacking Brackets model F420741500. 4.0 models can be converted from horizontal to vertical orientation by turning the unit and ordering vertical model shelves. Gas ports can be closed to use desiccator with standard desiccant cartridges models 420450250 or 420450200. Overall Dimensions without gas ports: 51.8W x 41.4D x 34.1 cmH (20.4 x 16.3 x 13.4").
Rear-mounted gas ports are tapered to accept tubing with ¼ to ⅜” I.D. The pull down “oven style” door has an easy-to-view digital hygrometer, a patented seal design, tab latches and security loops for a padlock or tamper-evident seals. Two removable, perforated shelves facilitate circulation of dry air.
Certifications: U.S. Patents: 6,834,920; 7,318,630 and D474,626.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty