Would you like to log in again?
OneRack® Tube Racks, Polypropylene (PP)
Supplier: Heathrow Scientific
OneRack®
HS27551A HS27552D HS27552B HS27552C HS27551B HS27551C HS27551D HS27552A
10147-563CS 102.68 USD InStock
10147-563 10147-561 10147-557 10147-567 10147-565 10147-559 10147-569 10147-571
OneRack® Tube Racks, Polypropylene (PP)
Racks Tube Racks
OneRack® Tube Racks are tough and highly resistant to acids, bases, solvents, and heat.
  • Made out of Polypropylene
  • Have molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
  • Unique tear drop shaped openings for better retention of tubes
  • Offers good chemical and heat resistance (0 to 110 °C)
Durable OneRack® Tube Racks are molded in a single, continuous piece so no assembly is required and there are no detachable pieces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty