Would you like to log in again?
durAstatic® Dissipative HDPE Bottles, Menda
Supplier: Menda
35737 35731 35734 35590 35498 35591 35497 35592 35263 35494 35261 35763 35736 35260 35733 35491 35264 35262 35495 35739 35499 35496 35593 35265 35492 35266 35493 35738 35764 35762 35732 35761
89499-048EA 10.26 USD InStock
89499-048 89499-004 89499-002 89499-046 10147-612 10147-610 89499-062 89499-060 89499-000 89499-066 89499-044 89499-064 89499-042 10147-624 10147-608 10147-586 10147-584 89499-058 10147-622 10147-620 89499-052 89499-050 89499-056 10147-582 89499-054 10147-580 89498-998 89498-996 89498-994 89498-992 10147-614 10147-618
durAstatic® Dissipative HDPE Bottles, Menda
Bottles Plastic Bottles
The dissipative HDPE durAstatic™ bottle is designed to protect valuable static sensitive printed circuitry.
  • Available in four colors: orange, green, blue, and yellow
  • No migratory additives
  • Embossed with ESD protective symbol
  • Colored bottles make it easy to identify various chemicals on the workstation
  • Printed with bottle contents to eliminate the need for additional labels
No migratory additives are used, to reduce the chance for contamination from the bottle. Colored bottles allow for quick and easy identification in the workstation.
Certifications: Long lasting dissipative properties: Rtt: 1×10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12; <2 second charge decay, tested per IEC 61340-5-1:1998.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty