Would you like to log in again?
durAstatic® Dissipative HDPE Bottles, with Black Screw Cap, Menda
Supplier: Menda
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
35818 35819 35820
10147-640EA 5.71 USD InStock
10147-640 10147-644 10147-642
durAstatic® Dissipative HDPE Bottles, with Black Screw Cap, Menda
Bottles Plastic Bottles
HDPE durAstatic® bottles are dissipative and are designed to protect valuable static sensitive printed circuitry.
  • No migratory additives
  • Embossed with ESD protective symbol
  • Vessel of 6 oz. includes measurements on one side to monitor fluid level
  • Black screw cap is insulative
Available in four colors for easy identification in the workplace, HDPE durAstatic® bottles are dissipative and are designed to protect valuable static sensitive printed circuitry. No migratory additives are used, reducing the chance for contamination from the bottle.
Black screw cap easily allows opening and reclosing to prevent contents from spilling. The cap is insulative.
Certifications: Long lasting dissipative properties: Rtt: 10⁴ to <1×10¹¹ ohms tested per ANSI/ESD STM11.12; <2 second charge decay, tested per IEC 61340-5-1:1998.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty