Would you like to log in again?
Static Dissipative Tool Roll-Up Pouches, Menda
Supplier: Menda
35981
10147-654EA 30.3 USD InStock
10147-654
Static Dissipative Tool Roll-Up Pouches, Menda
Tools Storage
Static dissipative tool roll-up pouches are ideal for employees to check out tools from tool cribs. They are perfect for organizing and issuing tools to operators.
  • Constructed of durable, blue vinyl
  • Rolls up for easy storage; hook and loop secures closure
  • Pockets designed to store up to 18 tools
  • Unrolled dimensions: 20×12.5"
Applications and industries: electricians, carpenters, contractors, maintenance, ironworkers, artisans, masonry. Perfect for organizing and issuing tools to operators. Rolls up for easy storage, with a hook and loop to secure closure.
Certifications: Rtt: 1×10⁴ to <1×10¹¹ ohms per ANSI/ESD S4.1
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty