Would you like to log in again?
Microwave Oven, Argos Technologies
Supplier: Argos Technologies
24406-40
10147-786EA 1016.34 USD InStock
10147-786
Microwave Oven, Argos Technologies
Ovens Microwave Ovens
The compact design makes this .7 cu foot microwave oven ideal for the crowded lab as it occupies minimal counter space
Its 700 watts of power prepares samples rapidly without overheating. The control panel is easy to read and operate, and can be locked to prevent changes to the program. The rapid heat feature sets the oven at high with a single touch. Internal glass turntable rotates during operation for even heat distribution.
Certifications: UL Listed.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty