Would you like to log in again?
Whatman™ Grade 113 Qualitative Filter Papers
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
1113-090 1113-185 1113-8768 1113-320 1113-500 1113-090 1113-500 1113-110 1113-125 1113-8768 1113-185 1113-917 1113-240 1113-150 1113-320
28297-258PK 51.02 USD InStock
28297-258 CA28297-254 CA28464-980 CA28464-990 28464-980 10147-908 CA76423-772 28464-990 CA28297-262 76423-772 28297-254 10147-910 10147-906 28297-262 28297-260
Whatman™ Grade 113 Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
Ultra high loading capacity with a particle retention well suited for use with coarse or gelatinous precipitates.
  • Fastest flow rate of the qualitative grades
  • Creped surface
  • Thickest filter paper in the qualitative range
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc. Some wet strengthened grades are available in folded (prepleated) forms.
This filter is also available in the FilterCup, a convenient, disposable 70 mm filter funnel with a 250 ml capacity, molded from polypropylene with an integral, heat bonded filter. Also available as Grade 113V.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty