Would you like to log in again?
Whatman™ Grade 230 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
5230-500 5230-400 5230-400 5230-090 5230-090 5230-330 5230-250 5230-150 5230-240 5230-500 5230-125 5230-150 5230-240 5230-240 5230-125 5230-110 5230-330 5230-185 5230-250 5230-110 5230-185 5230-200 5230-200
70240-122PK 38.35 USD InStock
70240-122 28319-280 CA70240-122 28297-337 28297-634 CA28319-360 28297-336 CA10147-950 28319-360 10147-950 CA28319-280 28297-630 CA28319-275 CA28297-630 28319-275 28319-304 CA28317-310 CA28297-336 CA28297-634 28317-310 CA28319-304 70240-118 CA70240-118
Whatman™ Grade 230 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A general qualitative filter with creped surface. Widely used in industrial laboratories and students laboratories for routine filtration.
  • Retains coarse particles
  • High speed and loading capacity
  • Strong, thick filter
The filter is wet-strengthened with a small amount of chemically stable resin to extend the life of the filter and improve handling. Normal qualitative applications will not introduce any significant impurities into the filtrate. However it is not recommended for Kjeldahl nitrogen analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty