Would you like to log in again?
GlycanPac AXR-1 Glycan Isomer Analysis Columns, Thermo Scientific
Supplier: Thermo Scientific
088136 088135
10148-524EA 1918.86 USD InStock
10148-524 10148-470
GlycanPac AXR-1 Glycan Isomer Analysis Columns, Thermo Scientific
Chromatography Columns
The Thermo Scientific™ GlycanPac™ AXR-1 column is a high-performance, silica-based HPLC column for the simultaneous separation of glycans by charge, isomerism, and size. It is designed to provide industry-leading resolution with unique selectivities for biologically important glycans, either labeled or native, using either fluorescence and/or mass spectrometry (MS) detection.
  • Unique glycan selectivity based on charge, branch/linkage isomerism, and size
  • Exceptional resolution for either native or derivatized glycans
  • Compatibility with both MS and fluorescence detection methods
The GlycanPac AXR-1 column combines both weak anion-exchange (WAX) and reversed-phase interaction mechanisms to produce exceptional glycan selectivity and resolution. The WAX functionality provides retention of negatively charged glycans that elute in order of increasing charge, and the reversed-phase mode facilitates resolution of glycans of the same charge according to their isomerism and size. The GlycanPac AXR-1 column is designed for HPLC and UHPLC methods using either fluorescence or MS detection, and uses volatile aqueous buffers (e.g., ammonium acetate or ammonium formate) and acetonitrile, presenting the eluting glycans ready for introduction into MS instruments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty