Would you like to log in again?
MAbPac™ HIC-10 Protein HIC Columns, Thermo Scientific
Supplier: Thermo Scientific
088481 088482 088480
10148-485EA 2011.13 USD InStock
10148-485 10148-487 10148-489
MAbPac™ HIC-10 Protein HIC Columns, Thermo Scientific
Chromatography Columns
The Thermo Scientific™ MAbPac™ HIC-10 column is a high resolution, silica-based HPLC column designed for separation monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction. The unique chemistry provides high resolution, excellent bio-compatibility, and selectivity that is different from other available HIC columns.
  • Unique chemistry designed for optimal selectivity for mAbs
  • Excellent bio-compatibility and extremely low carryover
  • High resolution and high efficiency
The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1000 Å), 5 μm silica particles functionalized with proprietary alkyl amide groups. The advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty