Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Glassvan® Miniature Blades, Sterile, MYCO Medical
Supplier: MYCO Medical
2002-67MIS 2002-61 2002-62 2002-64 2002-67
10148-794BX 37.24 USD InStock
10148-794 10148-782 10148-784 10148-830 10148-628
Glassvan® Miniature Blades, Sterile, MYCO Medical
Scalpels and Scalpel Assemblies Scalpel Blades
GLASSVAN® Miniature Blades are produced from high quality, high durability Swiss Carbon steel
Computerized grinding and proprietary polishing processes ensure consistent, clean, sharp cutting edges and two VCI strips ensure product and packaging integrity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty