Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Glassvan® Podiatry Blades, Sterile, MYCO Medical
Supplier: MYCO Medical
2003-81 2003-17 2003-84 2003-314 2003-88 2003-313 2003-312 2003-316 2003-09
10148-792BX 37.24 USD InStock
10148-792 10148-820 10148-854 10148-832 10148-802 10148-812 10148-804 10148-836 10148-828
Glassvan® Podiatry Blades, Sterile, MYCO Medical
Scalpels and Scalpel Assemblies Scalpel Blades
GLASSVAN® Podiatry Blades are produced from high quality, high durability Swiss Carbon and Stainless steel
Computerized grinding and proprietary polishing processes ensure consistent, clean, sharp cutting edges and two VCI strips ensure product and packaging integrity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty