Would you like to log in again?
Glassvan® Surgical Blades, Sterile, MYCO Medical
Supplier: MYCO Medical
2001T-25 2001T-10A 2001T-11 3001T-60 2001T-23 2001T-20 2001T-12 3001T-11 3001T-21 3001T-23 2001T-21 3001T-20 2001T-12B 3001T-22 3001T-15C 3001T-15 2001T-15C 3001T-12B 3001T-24 3001T-10 2001T-22 2001T-24 3001T-25 2001T-70GV 3001T-12 2001T-10 2001T-15
10148-818BX 111.93 USD InStock
10148-818 10148-810 10148-834 10148-814 10148-838 10148-816 10148-780 10148-840 10148-842 10148-786 10148-808 10148-800 10148-844 10148-822 10148-788 10148-846 10148-824 10148-826 10148-848 10148-806 10148-796 10148-630 10148-850 10148-798 10148-852 10148-632 10148-790
Glassvan® Surgical Blades, Sterile, MYCO Medical
Scalpels and Scalpel Assemblies Scalpel Blades
GLASSVAN® Surgical Blades are produced from high quality, high durability Swiss Carbon and Stainless steel
Computerized grinding and proprietary polishing processes ensure consistent, clean, sharp cutting edges and two VCI strips ensure product and packaging integrity.
Ordering information: Blade sizes #24 is special order item.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty