Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Chicken Polyclonal Antibody

Supplier: Immunoreagents

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CKXRT-003-D
10149-076EA 240.04 USD InStock
10149-076
Anti-IgG Chicken Polyclonal Antibody
Antibodies
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty