Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Donkey Polyclonal Antibody (Biotin (NHS-LC))

Supplier: Immunoreagents
DKXSH-003-GBIO
10149-432EA 231.17 USD InStock
10149-432
Anti-IgG Donkey Polyclonal Antibody (Biotin (NHS-LC))
Antibodies
ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin (NHS-LC)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty