Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Donkey Polyclonal Antibody (DyLight® 350)

Supplier: Immunoreagents
DKXSH-003-F350NHSX
10149-448EA 346.73 USD InStock
10149-448
Anti-IgG Donkey Polyclonal Antibody (DyLight® 350)
Antibodies
The ImmunoReagents' DyLight® 350 conjugates are a derivative of AMCA (Aminomethylcoumarine acetate), which a blue fluorescent dye commonly is used as a contrasting dye for double- and triple-labeling used in a wide array of applications. The UV-excitable DyLight® 350 has spectral properties that are very similar to those displayed by AMCA, including identical excitation wavelengths and a large stokes shift. The emission wavelength of DyLight® 350 has a 10-nm blue shift over AMCA, though, that provides an advantage in multiplexing with green fluorophores. (Excitation/Emission = 353nm / 432nm Emission Color = Blue (Similar Dyes: Alexa Fluor 350, AMCA)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 350
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty