Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (DyLight® 550)

Supplier: Immunoreagents
GTXMU-003-F550NHSX
10150-326EA 247.14 USD InStock
10150-326
Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Antibodies
The ImmunoReagents' DyLight® 550 conjugates provide vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives, including Alexa Fluor 555, TRITC and Cy3 for fluorescent applications. DyLight® 550 conjugates have tight excitation/emission spectra that are ideal for multicolor applications. DyLight® 550 conjugated secondary antibodies are used in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 562nm / 576nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, TRITC)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty