Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM µ Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Immunoreagents
RBXHU-006-DRHOX
10151-236EA 366.29 USD InStock
10151-236
Anti-IgM µ Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
TRITC (Tetramethylrhodamine isothiocyanate) is a popular rhodamine derivative pink in color but emits a red-orange color upon Emission at 580nm. TRITC is frequently paired with FITC in double labeling experiments and conjugated antibodies are often used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. These conjugates are best excited with the 532 nm spectral line of the He-Ne laser and are also recommended as a second color detection reagent in in situ hybridization applications. (Excitation/Emission = 555nm / 580nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, DyLight 550)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty