Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Immunoreagents
GTXHU-001-GFITC
10151-886EA 288.04 USD InStock
10151-886
Anti-IgA alpha Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
ImmunoReagents’ FITC Conjugates are used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. Fluorescein is typically excited by the 488 nm line of an argon laser, and emission is collected at 518 nm. FITC antibody conjugates are yellow-orange in color but emit a yellow-green color upon Em 518 nm. FITC antibody conjugates are sensitive to light. The typical FITC to antibody ratio (F/P) is >4.0 Excitation/Emission = 494nm / 518nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, DyLight ® 488)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: alpha chain
Host: Goat
Isotype: IgA
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty