Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Lama Polyclonal Antibody (DyLight® 488)

Supplier: Immunoreagents
LLXRB-003-D488NHSX
10152-182EA 172.47 USD InStock
10152-182
Anti-IgG Lama Polyclonal Antibody (DyLight® 488)
Antibodies
The ImmunoReagents' DyLight® 488 conjugates provide exceptional performance for a wide array of fluorescence-base applications at a lower cost than other 488-range dyes. DyLight® 488 shows greater photostability than traditional fluorescein-based dyes and has a greater range of pH insensitivity (pH 4-9). DyLight® 488 conjugates are available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 493nm / 518nm Emission Color = Green (Similar Dyes: Alexa Fluor 488, Cy2, FITC)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Llama
Isotype: IgG
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty