Would you like to log in again?
Polypropylene Cleanroom Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
NTP-811
10152-454CS 267.12 USD InStock
10152-454
Polypropylene Cleanroom Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These meltblown polypropylene wipes, embossed with a diamond pattern, are extremely low linting. They feature a high absorbency of 400% and are resistant to acids and other chemicals.
  • 100% polypropylene
  • Extremely absorbent
  • Chemically resistant to solvents
  • Flat and double-bagged
  • ESD safe pink pag
Compatible with ISO Class 5 Cleanroom. These polypropylene cleanroom wipes are used in a wide variety of industries like pharmaceutical, electonics, precision instruments and semiconducting.
Ordering information: Wipes are packaged 500 wipes per bag with 6 bags per case. Packaged in ESD safe pink bags.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty