Would you like to log in again?
Reagents with Lipid Clearing Factor (LCF) LiquiColor®, Stanbio Laboratory
Supplier: Stanbio Laboratory
LiquiColor® & Liqui-UV®
0590-080 0590-500 1010-430 2200-430 0710-080 2200-225 0590-040 0599-020 2100-430 2100-225 1010-225
10152-502EA 431.46 USD InStock
10152-502 10152-504 10152-506 10152-508 10152-570 10152-584 10152-572 10152-586 10152-574 10152-500 10152-576
Reagents with Lipid Clearing Factor (LCF) LiquiColor®, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
In some cases, patient specimens with increased lipid concentrations appear turbid
This turbidity can cause problems in the performance of photometric assays and can be avoided by employing pre-treatment steps. Such steps, however, are time-consuming and prone to error. Stanbio has developed a mixture of special additives, collectively called LCF, to eliminate these pre-treatment steps. LCF is integrated in the respective assay reagent and begins working immediately in the incubation phase of the test procedure. Clearing is achieved within minutes (e.g. up to 2000 mg/dL triglycerides, equivalent to 22.8 mmol/L, is cleared within 2-3 minutes at room temperature). Stanbio LCF assays, therefore, do not require additional pre-treatment steps allowing for easily obtained correct results.
Certifications: All lipids are FDA cleared and CE marked. Some lipids have additional Health Canada certifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty