Would you like to log in again?
Serology Kits, Stanbio Laboratory
Supplier: Stanbio Laboratory
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1130-100 1125-050 1130-050 1170-500 1155-050 1170-100 1125-100 1155-100
10152-612EA 90.45 USD InStock
10152-612 10152-622 10152-614 10152-624 10152-616 10152-618 10152-608 10152-610
Serology Kits, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
Our serology kits feature "no dilution" technology which eliminates the need to pre-dilute patient specimens
RaPET® kit includes Latex Reagent, Test Slide, Pipets/Mixers, Dilution Buffer and Controls. Results are Qualitative or Semi-Quantitative.
RPR Quicktest includes RPR Antigen Suspension, Pipets/Stirrers, Dispensing Needle, Dispensing Bottle, 18 mm Circle Test Cards and Three Levels of Controls. Results are Qualitative or Quantitative.
Stanbio Laboratory also offers a charcoal flocculation assay for detecting Syphilis (RPR).
Certifications: All serology tests are FDA cleared and CE marked. Some tests have additional Health Canada certifications.
Ordering information: The serology test kits comes complete with latex reagent, test slide, pipettes/mixers, dilution buffer and positive and negative controls.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty