Would you like to log in again?
Pregnancy Tests, Stanbio Laboratory
Supplier: Stanbio Laboratory
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1225-205 1440-050 1230-050 1220-025 1430-050 1220-050 1230-025 1235-305
10152-688EA 386.5 USD InStock
10152-688 10152-690 10152-692 10152-682 10152-696 10152-684 10152-698 10152-686
Pregnancy Tests, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
Stanbio Laboratory offers pregnancy tests to meet your clinical and laboratory needs.
Sensivity in Urine 20 mIU/mL
Sensivity in Serum 10 mIU/mL (20 mIU/mL for the True® 20 Plus set)
Certifications: All pregnancy tests are FDA cleared and CE marked. Some tests have additional Health Canada certifications.
Ordering information: Tests are single step cassette platform formats for urine only or urine/serum pregnancy testing.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty